Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ichthammol, USP, Spectrum™ Chemical
SDP

Catalog No. 18603413 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Drum
Poly Pail

Catalog No. 18-603-413

Supplier: Spectrum Chemical Manufacturing Corporation IC11012KG

Only null left
Add to Cart
Add to Cart

IC110, 8029-68-3

Spectrum™ Chemical Ichthammol, USP , also known as ammonium bituminosulfonate, is used to treat certain skin diseases like psoriasis and eczema. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8029-68-3

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 50%
Packaging Poly Pail
Quantity 12 kg
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.