Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ICI 182,780
GREENER_CHOICESDP

Catalog No. 104750 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-475-0 50 mg
10-471 1 mg
10-471-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 10-475-0 Supplier R&D Systems™ Supplier No. 1047/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Estrogen receptor antagonist

ICI 182,780 is a high affinity estrogen receptor antagonist (IC50 = 0.29 nM), devoid of any partial agonism both in vitro and in vivo. ICI 182,780 is a high affinity agonist at the membrane estrogen receptor GPER. ICI 182,780 inhibits MCF-7 human breast cancer cell growth in vitro and shows antitumor activity in mice models. ICI 182,780 penetrates the blood-brain-barrier and has neuronal functional effects in vivo.

Chemical Identifiers

CAS 129453-61-8
Molecular Formula C32H47F5O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 606.77
Purity >99%
Molecular Weight (g/mol) 606.777
Chemical Name or Material ICI 182, 780
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.