Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICI 199,441 hydrochloride, Tocris Bioscience™

Catalog No. 077810
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 07-781-0

Supplier: R&D Systems 0778/10

This item is not returnable. View return policy

Potent κ agonist

Highly potent κ agonist, 146-fold more active than U-50488 (Cat. No. 0495) in vitro.

Chemical Identifiers

CAS 115199-84-3
Molecular Formula C21H25Cl3N2O
Molecular Weight (g/mol) 427.794
InChI Key VFLWVWZSDBTGQJ-VEIFNGETSA-N
Synonym ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s
PubChem CID 3082717
IUPAC Name 2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride
SMILES CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 427.8
Percent Purity >99%
Chemical Name or Material ICI 199, 441 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.