Learn More
Tocris Bioscience™ ICI 89406
Description
Specifications
Specifications
| Chemical Name or Material | N-[2-[3-(2-Cyanophenoxy)-2-hydroxypropylamino]ethyl]-N'-phenylurea |
| CAS | 53671-71-9 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C19H22N4O3 |
| Formula Weight | Observed MW: 354.41 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.