Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ICI 89406
SDP

Catalog No. 083250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-325-0 50 mg
08-321-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 08-325-0 Supplier Tocris Bioscience™ Supplier No. 0832/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

β antagonist

β-adrenergic antagonist and low efficacy partial agonist; does not affect resting cardiac parameters.

Specifications

Chemical Name or Material N-[2-[3-(2-Cyanophenoxy)-2-hydroxypropylamino]ethyl]-N'-phenylurea
CAS 53671-71-9
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C19H22N4O3
Formula Weight Observed MW: 354.41

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.