Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287618
Description
ICK Polyclonal specifically detects ICK in Human samples. It is validated for Western Blot.Specifications
ICK | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
EC 2.7.11, hICK, intestinal cell (MAK-like) kinase, Intestinal cell kinase, KIAA0936EC 2.7.11.22, Laryngeal cancer kinase 2, LCK2serine/threonine protein kinase, MAK-related kinase, MGC46090, MRKECO, serine/threonine-protein kinase ICK | |
The immunogen is a synthetic peptide directed towards the C-terminal region of ICK | |
100 μL | |
Protein Kinase | |
22858 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction