Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113514
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113514 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113514 Supplier Invitrogen™ Supplier No. PA5113514
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ICK Polyclonal Antibody for Western Blot, IHC (P), ELISA

ICK or intestinal cell kinase belongs to a very extensive family of proteins that share a conserved catalytic core common with both serine/threonine and tyrosine protein kinases. ICK encodes an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein (MAP) kinases. ICK localizes to the intestinal crypt region and is thought to be important in intestinal epithelial cell proliferation and differentiation. ICK purified from transfected human embryonic kidney cells undergoes autophosphorylation and phosphorylates a test protein. ICK plays a key role in development of central nervous, skeletal, and endocrine systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CILK1
Gene Accession No. Q9UPZ9
Gene Alias 2210420N10Rik; AI848300; ciliogenesis associated kinase 1; CILK1; ECO; heart serine-threonine protein kinase; hICK; iciliogenesis associated kinase 1; ICK; intestinal cell (MAK-like) kinase; intestinal cell kinase; KIAA0936; Laryngeal cancer kinase 2; LCK2; MAK-related kinase; mICK; MRK; serine/threonine protein kinase; serine/threonine-protein kinase ICK
Gene Symbols CILK1
Host Species Rabbit
Immunogen Recombinant Human Serine/threonine-protein kinase ICK protein (150-275AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22858
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.