Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983859
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3859 Supplier Bioss Supplier No. BS15536R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Eukaryotic protein kinases are enzymes that belong to a very extensive family of proteins which share a conserved catalytic core common with both serine/threonine and tyrosine protein kinases. This gene encodes an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein kinases. The protein localizes to the intestinal crypt region and is thought to be important in intestinal epithelial cell proliferation and differentiation. Alternative splicing has been observed at this locus and two variants, encoding the same isoform, have been identified.

Specifications

Antigen ICK
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CILK1
Gene Accession No. Q9UPZ9
Gene Alias 2210420N10Rik; AI848300; BOS_21477; ECO; heart serine-threonine protein kinase; hICK; ICK; intestinal cell (MAK-like) kinase; Intestinal cell kinase; KIAA0936; Laryngeal cancer kinase 2; LCK2; MAK-related kinase; mICK; MRK; serine/threonine protein kinase; Serine/threonine-protein kinase ICK
Gene Symbols CILK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ICK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22858
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.