Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICMT Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297509UV
Description
ICMT Polyclonal specifically detects ICMT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ICMT | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
23463 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
EC 2.1.1.100, HSTE14, Isoprenylcysteine Carboxyl Methyltransferase, Isoprenylcysteine carboxylmethyltransferase, MST098, MSTP098, PCCMT, PCMT, PPMT, Prenylated Protein Carboxyl Methyltransferase, Protein-S-Isoprenylcysteine O-Methyltransferase | |
Produced in rabbits immunized with E. coli-derived Human ICMT fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction