Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICMT Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297509MFV450
Description
ICMT Polyclonal antibody specifically detects ICMT in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
ICMT | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
EC 2.1.1.100, HSTE14, Isoprenylcysteine Carboxyl Methyltransferase, Isoprenylcysteine carboxylmethyltransferase, MST098, MSTP098, PCCMT, PCMT, PPMT, Prenylated Protein Carboxyl Methyltransferase, Protein-S-Isoprenylcysteine O-Methyltransferase | |
Produced in rabbits immunized with E. coli-derived Human ICMT fragment. | |
0.1 mL | |
Cell Biology | |
23463 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction