Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICOS Antibody (2317C) [PE], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB1682P
Description
ICOS Monoclonal antibody specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOFSpecifications
ICOS | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
2317C | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
Chinese Hamster Ovary cell line, CHO-derived mouse ICOS, Met1-Leu142, Accession # Q9WVS0 | |
0.1 mL | |
Adaptive Immunity, Immunology | |
29851 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction