Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICOS Antibody (669214), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB69753E
Description
ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand InteractionSpecifications
ICOS | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Blockade of Receptor-ligand Interaction | |
669214 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction | |
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
NS0 mouse myeloma cell line transfected with human ICOS and mouse myeloma cell line NS0-derived recombinant human ICOS cocktail, Glu21-Phe141, Accession # Q9Y6W8 | |
0.1 mL | |
Adaptive Immunity, Immunology | |
29851 | |
Store at 4°C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction