Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICOS Antibody (669238) - Azide and BSA Free, R&D Systems™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | ICOS |
---|---|
Clone | 669238 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry, Blocking/Neutralizing, CyTOF-ready |
Applications | Flow Cytometry, Blocking Assay, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB69752SP
![]() |
Novus Biologicals
MAB69752SP |
25 μg |
Each for $206.50
|
|
|||||
MAB69752
![]() |
Novus Biologicals
MAB69752 |
100 μg |
Each for $499.50
|
|
|||||
Description
ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
ICOS | |
1.0 mg/mL | |
Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Immunology | |
PBS with No Preservative | |
29851 | |
Partial recombinant human ICOS protein (amino acids 21-141) [UniProt Q9Y6W8] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
669238 | |
Flow Cytometry, Blocking/Neutralizing, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
ICOS | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title