Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICOS Antibody (669238), PerCP, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB69752PCP
Description
ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry.Specifications
ICOS | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
ICOS | |
Partial recombinant human ICOS protein (amino acids 21-141) [UniProt Q9Y6W8] | |
100 μL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
669238 | |
Flow Cytometry | |
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
Mouse | |
Protein A or G purified | |
RUO | |
29851 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction