Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICOS (CD278) Armenian Hamster anti-Human, Mouse, Alexa Fluor™ 488, Clone: C398.4A, BD Phrmingen™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 569526
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human|Mouse samples. It is validated for Flow cytometry
Specifications
ICOS (CD278) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Armenian Hamster | |
Affinity Chromatography | |
RUO | |
29851 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG |
Flow Cytometry | |
C398.4A | |
Alexa Fluor 488 | |
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 | |
Mouse D10.G4.1 T-cell clone | |
25 μg | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction