Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS (CD278) Armenian Hamster anti-Human, Mouse, BV480, Clone: C398.4A, BD Horizon™

Catalog No. BDB566085 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566085 0.1 mg
BDB566087 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566085 Supplier BD Biosciences Supplier No. 566085

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Armenian Hamster Monoclonal Antibody

ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human|Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICOS (CD278)
Applications Flow Cytometry
Classification Monoclonal
Clone C398.4A
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 480
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115
Host Species Armenian Hamster
Immunogen Mouse D10.G4.1 T-cell clone
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.