Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS (CD278) Armenian Hamster, anti-Human, R718, Clone: C398.4A, BD Horizon™

Catalog No. BDB566990 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566990 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566990 Supplier BD Biosciences Supplier No. 566990
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The C398.4A monoclonal antibody specifically binds to Inducible Costimulator (ICOS), which is also known as, CD278, Activation-inducible lymphocyte immunomediatory molecule (AILIM), or H4. ICOS is a type I transmembrane glycoprotein that forms a disulfide-linked homodimer and belongs to the CD28 family within the Ig superfamily. ICOS is expressed on either CD4+ or CD8+ single-positive mature thymocytes, T cells, or subsets of Innate Lymphoid Cells (ILC). Its expression is upregulated on activated T lymphocytes. ICOS is a costimulatory receptor that can bind to the B7-H2 ligand (CD275, ICOS-L) that is expressed on B cells, monocytes, macrophages, dendritic cells, and endothelial cells. ICOS plays a critical role in many types of T cell-dependent immunity. In the case of humoral immunity, for example, ICOS signaling is critical for the differentiation of T follicular helper (Tfh) cells and development of germinal centers. Although C398.4A was generated against mouse ICOS, this antibody reportedly crossreacts with human, rhesus, and rat ICOS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ICOS (CD278)
Applications Flow Cytometry
Classification Monoclonal
Clone C398.4A
Conjugate R718
Host Species Armenian Hamster
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.