Learn More
ICOS (CD278) Armenian Hamster, BUV395, Clone: C398.4A , BD Horizon™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | ICOS (CD278) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | C398.4A |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Ultraviolet 395 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 |
| Host Species | Armenian Hamster |
| Immunogen | Mouse D10.G4.1 T-cell clone |
| Show More |
Safety and Handling
For Reseach Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.