Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICOS (CD278) Armenian Hamster, BUV395, Clone: C398.4A, BD Horizon™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 565884
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
ICOS (CD278) | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Armenian Hamster | |
0.1 mg | |
Cell Biology | |
Human, Mouse, Rat, Rhesus Monkey | |
Purified |
Flow Cytometry | |
C398.4A | |
Brilliant Ultraviolet 395 | |
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 | |
Mouse D10.G4.1 T-cell clone | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions | |
IgG |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction