Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ICOS/CD278, Armenian Hamster, Unlabeled, Clone: C398.4A,

Catalog No. MABF977MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF977MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF977MI Supplier MilliporeSigma™ Supplier No. MABF977
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Inducible T-cell costimulator (UniProt Q9WVS0; also known as Activation-inducible lymphocyte immunomediatory molecule, CCLP, CD278, CD28 and CTLA-4-like protein, CD28-related protein 1, CRP-1, H4) is encoded by the Icos (also known as Ailim) gene (Gene ID 54167) in murine species. ICOS is a costimulatory molecule expressed on activated T cells and regulatory T cells, it plays a crucial role in T-cell survival and function, Th2 cell differentiation, and lung inflammatory responses. Upon ICOS-ligand (ICOS-L) binding, ICOS initiates a cascade of intracellular signaling events, leading to the production of cytokines such as IL-4 and IL-13. Both ICOS and ICOS-L are expressed on type 2 innate lymphoid cells (ILC2s) that, along with Th2 cells, contribute to the pathogenesis of asthma by producing copious amounts of IL-5 and IL-13. ICOS and ICOS-L interaction is reported to promote cytokine production and survival of ILC2s through STAT5 signaling. ICOS-deficiency or blocking ICOS and ICOS-L interaction is shown to prevent cytokine-induced asthma and lung inflammation in a humanized murine model with adoptively transferred human peripheral ILC2s. Human and murine ICOS proteins are produced with a signal peptide (a.a. 1-20) sequence, which is removed posttranslationally to yield the mature protein with a large N-terminal extracellular (human a.a. 21-140 and murine a.a. 21-144) region composed almost entirely of a V-type Ig-like domain (human a.a. 30-132 and murine a.a. 30-133), followed by a transmembrane domain (human a.a. 141-161 and murine a.a. 145-165) and a cytoplasmic tail (human a.a. 162-199 and murine a.a. 166-200).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICOS/CD278
Applications Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone C398.4A
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified Armenian hamster monoclonal antibody IgG in PBS without preservatives.
Gene Icos, Ailim
Gene Accession No. Q9WVS0
Host Species Armenian Hamster
Immunogen D10.G4.1 murine TH2 cells (Redoglia, V., et al. (1996). Eur. J. Immunol. 26(11):2781-2789).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_059508
Test Specificity Clone C398.4A recognizes a extracellular epitope present in murine (UniProt Q9WVS0) and the two alternatively spliced human ICOS/CD278 isoforms (UniProt Q9Y6W8).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.