Learn More
ICOS (CD278) Mouse anti-Human , Alexa Fluor 488, Clone: DX29, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | ICOS (CD278) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DX29 |
| Conjugate | Alexa Fluor 488 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | ICOS; DX-29; H4; Inducible T-cell costimulator; AILIM; CVID1 |
| Host Species | Mouse |
| Immunogen | Activated human T cells |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.