Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICP27 Rabbit anti-Virus, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 101861AP150UL
Description
There are two serotypes of the herpes simplex virus, HSV-1 (also known as type 1 or oral) and HSV-2 (type 2 or genital), can establish lifelong latent infections within sensory ganglia. Periodically, the virus reactivates and can cause re-current cold sores, eye and genital infections and encephalitis. One of the HSV-1/2 proteins involved in converting the cell into an efficient producer of viral gene products is the Infected Cell Polypeptide 27 or ICP27. HSV-1/2 immediate-early protein ICP27 is a nuclear phosphoprotein that is required for viral growth during lytic infection. Analysis of viral mutants defective in this function has shown that ICP27 has a number of effects on gene expression including a contribution to the shut off of host protein synthesis, the stimulation of HSV-1/2 early gene expression and DNA replication, and the induction of late viral gene products. ICP27 performs these functions primarily post-transcriptionally at the level of RNA processing. ICP27 affects three important RNA processing events: polyadenylation, splicing and nuclear RNA export.Specifications
| ICP27 | |
| Polyclonal | |
| Unconjugated | |
| ICP27, Immediate-early protein IE63, Infected cell protein 27, mRNA export factor, UL54, VMW63 | |
| ICP27 Fusion Protein Ag0239 | |
| 150 μL | |
| Primary | |
| -20°C | |
| Liquid |
| ELISA | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction