Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICSA Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144412 Supplier Invitrogen™ Supplier No. PA5144412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ICSA Polyclonal Antibody for Western Blot, ELISA

An immunogenic protein essential for bacterial spreading, it elicits polar deposition of filamentous actin (actin-based motility) (PubMed:2542950, PubMed:8702989). Inside the host (human) cell mediates nucleation and polymerization of actin molecules on the bacterial surface, which provides the propulsive force for intracellular movement and intercellular dissemination of the bacterium (PubMed:2644195, PubMed:8468279, PubMed:2542950). During invasion of mammalian cells, triggers autophagy by binding to host ATG5. Interaction with IcsB leads to escape from the host autophagy defense system (PubMed:15576571). Binds ATP and displays weak ATPase activity (PubMed:8468279). Mediates polar adhesion to host cells and their invasion. The bile salt deoxycholate activates adhesion and invasion of host cells, possibly by altering IcsA's conformation. The adhesion and intercellular dissemination phenotypes can be separated (PubMed:24721572).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICSA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.745 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene icsA
Gene Accession No. Q7BCK4
Gene Alias virG
Gene Symbols icsA
Host Species Rabbit
Immunogen Recombinant Shigella flexneri ICSA protein (311-547aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1238021
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.