Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICT Antibody (1E9), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242633AF594
Description
ICT Monoclonal specifically detects ICT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
ICT | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MRPL58 | |
Recombinant human ICT1 (30-206aa) purified from E. coli | |
Cancer | |
3396 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA | |
1000000000 | |
Western Blot, Flow Cytometry, ELISA | |
Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial | |
Mouse | |
100 μL | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction