Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICT Antibody (1E9), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242633MFV500
Description
ICT Monoclonal antibody specifically detects ICT in Human samples. It is validated for Flow CytometrySpecifications
ICT | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1000000000 | |
Flow Cytometry | |
Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial | |
Recombinant human ICT (30-206aa) purified from E. coli | |
0.1 mL | |
Cancer | |
3396 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction