Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICT Antibody (1E9), PerCP, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242633PCP

Catalog No. NBP242633JP


Only null left
Add to Cart

Description

Description

ICT Monoclonal specifically detects ICT in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ICT
Monoclonal
PerCP
PBS with 0.05% Sodium Azide
MRPL58
Recombinant human ICT1 (30-206aa) purified from E. coli
100 μL
Cancer
3396
Store at 4C in the dark.
IgG1 κ
Flow Cytometry, ELISA
1000000000
Flow Cytometry
Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.