Learn More
ICT1 Mouse anti-Human, Clone: OTI2F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505881

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Essential peptidyl-tRNA hydrolase component of the mitochondrial large ribosomal subunit. Acts as a codon-independent translation release factor that has lost all stop codon specificity and directs the termination of translation in mitochondrion, possibly in case of abortive elongation. May be involved in the hydrolysis of peptidyl-tRNAs that have been prematurely terminated and thus in the recycling of stalled mitochondrial ribosomes.Specifications
ICT1 | |
Monoclonal | |
Unconjugated | |
MRPL58 | |
DS-1, DS1, ICT1, MRP-L58 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3396 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14197 | |
MRPL58 | |
Full length human recombit protein of human ICT1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.