Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551980 Supplier Invitrogen™ Supplier No. PA551980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEFKSIYSLD KLYPESQGSD TAWRVPNGAK QADSDIPLDR LTISYCRSSG PGGQNVNKVN SKAEVRFHLA TAEWIAEPVR QKIAITHKNK INRLGELILT SESSRYQFRN LADCLQKIRD MI Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

Essential peptidyl-tRNA hydrolase component of the mitochondrial large ribosomal subunit. Acts as a codon-independent translation release factor that has lost all stop codon specificity and directs the termination of translation in mitochondrion, possibly in case of abortive elongation. May be involved in the hydrolysis of peptidyl-tRNAs that have been prematurely terminated and thus in the recycling of stalled mitochondrial ribosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL58
Gene Accession No. Q14197
Gene Alias 1110001A02Rik; 1110002E03Rik; 39S ribosomal protein L58, mitochondrial; Digestion substraction 1; DS1; DS-1; Ict1; immature colon carcinoma transcript 1; immature colon carcinoma transcript 1 protein; Immature colon carcinoma transcript 1 protein homolog; Mitochondrial large ribosomal subunit protein mL62; mitochondrial ribosomal protein L58; MRPL58; MRP-L58; Peptidyl-tRNA hydrolase ICT1, mitochondrial
Gene Symbols MRPL58
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ICT1. Recombinant protein control fragment (Product #RP-101896).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3396
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.