Learn More
ICT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 104031AP150UL
Description
This antibody is a rabbit polyclonal antibody raised against a full-length human ICT1 protein.
Essential peptidyl-tRNA hydrolase component of the mitochondrial large ribosomal subunit. Acts as a codon-independent translation release factor that has lost all stop codon specificity and directs the termination of translation in mitochondrion, possibly in case of abortive elongation. May be involved in the hydrolysis of peptidyl-tRNAs that have been prematurely terminated and thus in the recycling of stalled mitochondrial ribosomes.Specifications
| ICT1 | |
| Polyclonal | |
| Unconjugated | |
| MRPL58 | |
| Digestion substraction 1, DS 1, DS1, ICT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303673, 3396, 68572 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14197, Q8R035 | |
| MRPL58 | |
| ICT1 Fusion Protein Ag0647 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.