Learn More
QuidelOrtho ID-Micro Typing System™ Gel Test™ Cards
SDP
Catalog No. OKMTS080312
Click to view available options
Quantity:
100 Cards
20 Cards
Type:
ID-Micro Typing System™ A/B Monoclonal Grouping Card
ID-Micro Typing System™ A/B/D Monoclonal Grouping Card
ID-Micro Typing System™ A/B/D Monoclonal and Reverse Grouping Card
ID-Micro Typing System™ Anti-IgG,-C3d Card
ID-Micro Typing System™ Buffered Gel Card
ID-Micro Typing System™ MTS Anti-IgG Card
ID-Micro Typing System™ MTS Anti-c (Monoclonal) Card
ID-Micro Typing System™ MTS Anti-e (Monoclonal) Card
ID-Micro Typing System™ MTS Monoclonal Control Card
ID-Micro Typing System™ MTS Monoclonal Rh Phenotype Card
ID-Micro Typing System™ MTS anti-A (Murine Monoclonal) card
ID-Micro Typing System™ MTS anti-B (Murine Monoclonal) card
This item is not returnable. View return policy
This item is not returnable. View return policy
Delivered with gel-filled microtubes infused with pre-measured reagents
QuidelOrtho ID-Micro Typing System is suitable for a broad range of blood bank applications, including antibody screening and identification, ABO blood grouping and Rh phenotyping, compatibility testing, and reverse serum grouping and antigen typing, all using proven serologic methods. Gel Test cards are prefilled cards containing gel mixed with the appropriate reagent. Simply pipet, incubate, spin and read: results are ready to report, without the need for cell washing, red cell resuspension or tube shaking.
Standardization
  • Standardized methods ensure uniform results from tech-to-tech and laboratory-to-laboratory
  • Enhanced cGMP compliance is attained through standardized procedures and simplified training
Safety
  • Opportunities for error are reduced by stable and clear endpoints that provide objective, consistent results
  • Ease of Use
  • Gel testing is easy to learn, use, and teach: the ideal technology for all shifts and all levels of blood banking experience
  • Eliminates washing, shaking, and the need to add control cells
Cost-effectiveness
  • Gel testing requires fewer steps than tube testing, dramatically reducing hands-on time, with less waste and costly re-work
  • Well-balanced sensitivity and specificity means less time and fewer resources spent on clinically unimportant antibodies
Specifications
Certifications/Compliance cGMP
Type ID-Micro Typing System™ MTS Anti-e (Monoclonal) Card
For Use With (Application) For broad range of Blood bank applications, Antibody screening and identification, ABO Blood grouping and Rh phenotyping, compatibility testing and reverse Serum grouping and Antigen typing
Includes Gel-filled microtubes
Quantity 20 Cards
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.