Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ ID NOW™ COVID-19 Assay Kit

Catalog No. 23046650 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
24 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-650 24 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-650 Supplier Abbott™ Supplier No. 190000
This item is not returnable. View return policy

High-quality molecular technology targets the COVID-19 RdRp gene for a positive result within 5 minutes, and a negative result within 13 minutes.

  • Leading molecular point-of-care (POC) platform in the United States
  • Trusted by hospitals, physician offices, and urgent care facilities nationwide
  • Emergency Use Authorization (EUA) supports flexible near-patient testing environments
  • Sample types tested direct or in viral transport media (VTM) include throat swabs, nasal swabs, and nasopharyngeal swabs
  • Room temperature storage eliminates the need for refrigeration
  • Assay kit includes 24 tests, sample collection swabs, pipettes, and control swabs
  • Control kit is available for addition positive and negative swabs

Specifications

Certifications/Compliance CLIA
Content And Storage Ambient
Control Sets 12 Positive and 12 Negative (Additional Controls Sold Separately)
CE Marker Yes
Clia Complexity Waived, Moderate, High
Detectable Analytes COVID-19
Format Kit
Sample Type Nasal Swabs, Nasopharyngeal Swabs, and Throat Swabs
Sensitivity See Package Insert
Type Molecular COVID Assay
Disposable Single-use
For Use With (Application) Isothermal Molecular
For Use With (Equipment) ID NOW™ Instrument
Includes 24 Tests, Sample Collection Swabs, Pipettes, and Control Swabs
Material Plastic
Organism Group Virus
Packaging Type 24/Kit
Quantity 24 Tests
Specificity See Package Insert
Target Gene RdRp
Target Organism Class Virus
Test Time 5 Minutes (Positive), 13 Minutes (Negative)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.