Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ ID NOW™ Influenza A and B 2 PROMO

Catalog No. 23046634PM Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Detectable Analytes: Influenza A and B
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-046-634PM Detectable Analytes: Influenza A and B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-634PM Supplier Abbott™ Supplier No. 427000P

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The fastest molecular flu test just got faster and even better. Now with early detection and positive results in as few as 5 minutes, allowing you to make effective clinical decisions sooner. ID NOW™ is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology. Buy 5, get 6.

Transforming near-patient testing:
  • Unique isothermal amplification technology giving you molecular results, faster than ever before.
Improving patient outcomes:
  • Giving you the confidence to make effective clinical decisions sooner.
Optimizing operational efficiency:
  • Enables prompt initiation of infection control measures.
  • Connected results direct to your network.
  • Facilitating effective patient management.
Enabling effective Antimicrobial Stewardship:
  • Molecular sensitivity in an actionable time-frame allows the prudent use of antibiotics and antivirals.
A positive impact on your facility:
  • Reliable near-patient testing reduces overall healthcare costs.
Confidence at the point of care:
  • Intuitive and reliable, allowing you to respond quickly and accurately in any setting.
  • Simple and easy to use molecular diagnostic test.
  • Sample type nasal swab.
  • Requires only minimal training.
  • Small footprint can be used in any healthcare setting.
  • Streamlined workflow connectivity can be provided to your network.
  • Demonstrates impressive sensitivity for the detection of both influenza A and B in clinical respiratory specimens.

Specifications

Certifications/Compliance CLIA waived
Content And Storage Room Temperature
Description Detects up to 42% more true positives than RADTs
Detectable Analytes Influenza A and B
Type Abbott ID NOW Influenza A and B Test Kit
For Use With (Application) Influenza testing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.