Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ ID NOW™ RSV Test Kit PROMO

Catalog No. 23043632PM Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
24 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-632PM 24 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Refer to Package Insert for list of warnings
Catalog No. 23-043-632PM Supplier Abbott™ Supplier No. 435000P

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Refer to Package Insert for list of warnings

Highly sensitive rapid diagnostic tests for RSV are needed to help guide management in the clinical setting. ID NOW™ RSV Test Kit combines the benefits of speed and accuracy on an easy to use system. Buy 5, get 6.

  • Rapid molecular in vitro diagnostic test utilizing isothermal Nucleic Acid Amplification Technology for the qualitative detection of Respiratory Syncytial Virus l nucleic acid in nasopharyngeal swabs or nasopharyngeal swabs
  • Eluted in Viral Transport Media (VTM)
  • Intended to aid in the rapid diagnosis of Respiratory Syncytial Virus (RSV)
  • Delivers molecular RSV results in 13 minutes or less on the unique Abbott™ ID NOW platform.
  • Allows to make real-time clinical decisions which impact patient care
  • Includes Test Bases, Sample Receivers, Transfer Cartridges, NP Swabs, Positive Control Swab, Negative Control Swab, Plastic Disposable Pipettes capable of delivering 200 μL VTM sample, Package Insert, Quick Reference Instructions

Certifications

CLIA-waived

Specifications

Certifications/Compliance CLIA waived
Content And Storage Room Temperature
CPT Code 87634-QW
CE Marker Yes
Clia Complexity Waived
Detectable Analytes Respiratory Syncytial Virus
Sample Type Nasopharyngeal Swab, Nasopharyngeal Swab Eluted in Viral Transport Media (VTM)
Sensitivity Direct NPS: 98.6%, NPS eluted in VTM: 98.6%
Type Test Kit
For Use With (Application) For diagnosis of Respiratory Syncytial Virus (RSV)
Includes Test Bases, Sample Receivers, Transfer Cartridges, Swabs, Plastic Disposable Pipettes, Package Insert, Quick Reference Instructions
Material Cardboard
Packaging Type Cardboard Box
Quantity 24 Pk.
Specificity Direct NPS: 98%, NPS eluted in VTM: 97.8%
State at Room Temperature Box 1: Powder, Box 2: Liquid
Test Time 13 min. or less
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.