Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ID1 Antibody (PCRP-ID1-2F11), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314075JF646
This item is not returnable.
View return policy
Description
ID1 Monoclonal antibody specifically detects ID1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| ID1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| BHLHB24, bHLHb24DNA-binding protein inhibitor ID-1, Class B basic helix-loop-helix protein 24, dJ857M17.1.2, dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helixprotein), ID, inhibitor of differentiation 1, Inhibitor of DNA binding 1, inhibitor of DNA binding 1, dominant negative helix-loop-helix protein | |
| Recombinant fragment (aa 5-150) of human ID1 (Uniprot: P41134) | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3397 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-ID1-2F11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction