Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ID1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121198 Supplier Invitrogen™ Supplier No. PA5121198
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Rat heart. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MKVASGSTAT AAAGPSCALK AGKTASGAGE VVRCLSEQSV AISRCAGGAG ARLPALLDEQ QVNVLLYDMN GCYSRLKELV PTLPQNRKVS KVEILQHVID YIRDLQLELN SESEVGTPGG RGLPVRAPLS TLNGEISALT AEAACVPADD RILCR.

ID1 is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. It has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene.The protein encoded by this gene is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. The encoded protein has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ID1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ID1
Gene Accession No. P20067, P41134, P41135
Gene Alias AI323524; bHLHb24; class B basic helix-loop-helix protein 24; D2Wsu140e; dJ857M17.1.2; dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helix protein); DNA-binding protein inhibitor ID-1; ID; ID1; Id-1; ID125A; Idb1; inhibitor of differentiation 1; Inhibitor of DNA binding 1; Inhibitor of DNA binding 1 helix-loop-helix protein (splice varaiation); inhibitor of DNA binding 1, dominant negative helix-loop-helix protein; Inhibitor of DNA binding 1, helix-loop-helix protein (splice variation); inhibitor of DNA binding 1, HLH protein
Gene Symbols ID1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-155 of human ID1 (NP_0021562).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15901, 25261, 3397
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.