Learn More
Invitrogen™ ID1 Recombinant Rabbit Monoclonal Antibody (JM92-13)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA532749
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
ID1 is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. It has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene.The protein encoded by this gene is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. The encoded protein has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
ID1 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P41134 | |
ID1 | |
Synthetic peptide within Human Id1 aa 11-60. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
JM92-13 | |
Unconjugated | |
ID1 | |
AI323524; bHLHb24; class B basic helix-loop-helix protein 24; D2Wsu140e; dJ857M17.1.2; dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helix protein); DNA-binding protein inhibitor ID-1; ID; ID1; Id-1; ID125A; Idb1; inhibitor of differentiation 1; Inhibitor of DNA binding 1; Inhibitor of DNA binding 1 helix-loop-helix protein (splice varaiation); inhibitor of DNA binding 1, dominant negative helix-loop-helix protein; Inhibitor of DNA binding 1, helix-loop-helix protein (splice variation); inhibitor of DNA binding 1, HLH protein | |
Rabbit | |
Protein A | |
RUO | |
3397 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.