Learn More
ID2 Mouse anti-Human, Clone: OTI10C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500183

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the inhibitor of DNA binding family, members of which are transcriptional regulators that contain a helix-loop-helix (HLH) domain but not a basic domain. Members of the inhibitor of DNA binding family inhibit the functions of basic helix-loop-helix transcription factors in a domit-negative manner by suppressing their heterodimerization partners through the HLH domains. This protein may play a role in negatively regulating cell differentiation. A pseudogene of this gene is located on chromosome 3.Specifications
ID2 | |
Monoclonal | |
Unconjugated | |
ID2 | |
GIG8, ID2A, ID2H, bHLHb26 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3398 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI10C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q02363 | |
ID2 | |
Full length human recombit protein of human ID2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.