Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985461
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5461 Supplier Bioss Supplier No. BS3515R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The inhibitor of DNA binding 2 (ID2) protein is essential for promoting the development of a number of cell types including natural killer cell, erythroid precursors, as well as, CD8+ and CD4+ T-cell. A lack of ID2 promotes B-cell development. The chief role of this protein is to associate with ubiquitously-expressed E proteins, and other proteins of the basic helix-loop-helix (HLH) family, preventing them from binding to DNA and activating transcription. Activation of ID2 is critical for the development of the natural killer cell lineage.

Specifications

Antigen ID2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ID2
Gene Accession No. Q02363
Gene Alias Ac2-300; AI255428; BHLHB26; C78922; cell growth-inhibiting gene 8; Class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID2; DNA-binding protein inhibitor ID-2; GIG8; helix-loop-helix protein ID2; ID2; Id-2; ID2A; ID2H; Idb2; Inhibitor of differentiation 2; Inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein; inhibitor of DNA binding 2, HLH protein; MGC26389; OTTHUMP00000140258; OTTHUMP00000200297
Gene Symbols ID2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GIG8/ID2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3398
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.