Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ID3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555152 Supplier Invitrogen™ Supplier No. PA555152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPRGTQLSQV EILQRVIDYI LDLQVVLAEP APGPPDGPHL PIQTAELAPE LVISND Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Members of the Id family of basic helix-loop-helix (bHLH) proteins include ID1, ID2, ID3 and ID4. They are ubiquitously expressed and dimerize with members of the class A and B HLH proteins. Due to the absence of the basic region, the resulting heterodimers cannot bind DNA. The ID-type proteins thus appear to negatively regulate DNA binding of bHLH proteins. Since ID1 inhibits DNA binding of E12 and Myo D, it apparently functions to inhibit muscle-specific gene expression. Under conditions that facilitate muscle cell differentiation, the ID protein levels fall, allowing E12 and/or E47 to form heterodimers with Myo D and myogenin, which in turn activate myogenic differentiation. It has been shown that expression of each of the ID proteins is strongly dependent on growth factor activation and that reduction of ID mRNA levels by antisense oligonucleotides leads to a delayed reentry of arrested cells into the cell cycle following growth factor stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ID3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Id3
Gene Accession No. Q02535
Gene Alias 1R21; bHLHb25; class B basic helix-loop-helix protein 25; DNA-binding protein inhibitor ID-3; HEIR 1; HEIR1; HEIR-1; Helix-loop-helix protein HEIR-1; Hlh462; HLH642; ID 3; Id3; Id-3; Idb3; ID-like protein inhibitor HLH 1R21; ID-like protein inhibitor HLH 462; Inhibitor of differentiation 3; inhibitor of DNA binding 3; inhibitor of DNA binding 3, dominant negative helix-loop-helix protein; inhibitor of DNA binding 3, HLH protein
Gene Symbols Id3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ID3. Recombinant protein control fragment (Product #RP-94113).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3399
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.