Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ID4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP25632225UL
Description
ID4 Polyclonal specifically detects ID4 in Human, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ID4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein | |
Rabbit | |
Affinity Purified | |
RUO | |
3400 | |
Human, Rat | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
ID4 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PLTALNTDPAGAVNKQGDSILCR | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction