Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ID4 Antibody (PCRP-ID4-1A1), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | ID4 |
---|---|
Clone | PCRP-ID4-1A1 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 μg/mL, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate |
Applications | Western Blot, ELISA, Immunoprecipitation |
Description
ID4 Monoclonal antibody specifically detects ID4 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
ID4 | |
0.2 mg/ml | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein | |
Recombinant fragment (around aa 62-112) of human ID4 protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
PCRP-ID4-1A1 | |
Western Blot 1-2 μg/mL, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS with 0.05% BSA | |
3400 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title