Learn More
Invitrogen™ ID4 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ID4.
Specifications
Specifications
| Antigen | ID4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Id4 |
| Gene Accession No. | P41139, P47928 |
| Gene Alias | bHLHb27; cb547; Class B basic helix-loop-helix protein 27; DNA-binding protein inhibitor ID-4; fb77d04; hypothetical protein LOC664761; ID4; Id-4; Idb4; Inhibitor of differentiation 4; inhibitor of DNA binding 4; inhibitor of DNA binding 4, dominant negative helix-loop-helix protein; inhibitor of DNA binding 4, HLH protein; sb:cb547; wu:fb77d04; zgc:123214 |
| Gene Symbols | Id4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.