Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ID4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566347 Supplier Invitrogen™ Supplier No. PA566347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLTALNTDPAG AVNKQGDSIL CR Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

ID4 is transcription factors containing a basic helix-loop-helix (bHLH) motif regulate expression of tissue-specific genes in a number of mammalian and insect systems. DNA-binding activity of the bHLH proteins is dependent on formation of homo- and/or heterodimers. Dominant-negative HLH proteins encoded by Id-related genes, such as ID4, also contain the HLH-dimerization domain but lack the DNA-binding basic domain. Consequently, Id proteins inhibit binding to DNA and transcriptional transactivation by heterodimerization with bHLH proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ID4
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Id4
Gene Accession No. P47928
Gene Alias bHLHb27; cb547; Class B basic helix-loop-helix protein 27; DNA-binding protein inhibitor ID-4; fb77d04; hypothetical protein LOC664761; ID4; Id-4; Idb4; Inhibitor of differentiation 4; inhibitor of DNA binding 4; inhibitor of DNA binding 4, dominant negative helix-loop-helix protein; inhibitor of DNA binding 4, HLH protein; sb:cb547; wu:fb77d04; zgc:123214
Gene Symbols Id4
Host Species Rabbit
Immunogen Recombinant Human ID4. Recombinant protein control fragment (Product #RP-107030).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.