Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ID4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
| Antigen | ID4 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ID4 Polyclonal antibody specifically detects ID4 in Human samples. It is validated for ImmunofluorescenceSpecifications
| ID4 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PLTALNTDPAGAVNKQGDSILCR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 3400 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title