Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Idarucizumab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541998
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541998 Supplier Invitrogen™ Supplier No. MA541998
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Idarucizumab is a humanized monoclonal antibody fragment (Fab) derived from an immunoglobulin G1 isotype molecule that binds to and inactivates the oral anticoagulant dabigatran, thereby reversing its anticoagulant effect. As a direct acting oral anticoagulant (DOAC), one of the risks associated with the use of dabigatran includes bleeding, especially when given to patients at increased risk (elderly, chronic kidney disease, concomitant NSAID or warfarin use, etc.). Approved under the tradename Praxbind (FDA), idarucizumab is indicated for the emergency treatment of dabigatran-associated bleeding in life-threatening or surgically induced situations. Its use is associated with immediate, complete and sustained reversal of the anticoagulant effects of dabigatran. Idarucizumab protein structure can be viewed below, with disulfide bridges at the following points: H22-H95, H149-H205, H225-L-219, L23-L93, L139-L199.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Idarucizumab Humanized
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 2.53 mg/mL
Conjugate Unconjugated
Formulation 30mM histidine, PBS with 10% sucrose, 60ppm Tween 80 and no preservative; pH 6.0
Gene Alias aDabi-Fab; dabigatran; dabigatran etexilate mesylate
Host Species Human
Immunogen Pradaxa (Dabigatran Etexilate Mesylate) dabigatran etexilate mesylate.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 F(ab) κ
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.