Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Idasanutlin
GREENER_CHOICE

Catalog No. 690410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-041-0 10 mg
69-045-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-041-0 Supplier Tocris Bioscience™ Supplier No. 6904/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent MDM2 inhibitor; inhibits MDM2-p53 interaction

Idasanutlin is a potent MDM2 inhibitor (IC50 = 6 nM in binding assay, and 30 nM in cancer cell proliferation assay). Induces p53 stabilisation, cell cycle arrest and apoptosis in cancer cells expressing wildtype p53. Displays inhibition of tumor growth in the SJSA1 tumor xenograft model. Also inhibits MDR-1 at high concentrations.

Specifications

Quantity 10 mg
Molecular Formula C31H29Cl2F2N3O4
Formula Weight 616.49
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.