Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDE Recombinant Rabbit Monoclonal Antibody (8O8F3)
GREENER_CHOICE

Catalog No. PIMA535075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535075 Supplier Invitrogen™ Supplier No. MA535075
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MRYRLAWLLH PALPSTFRSV LGARLPPPER LCGFQKKTYS KMNNPAIKRI GNHITKSPED KREYRGLELA NGIKVLLISD PTTDKSSAAL DVHIGSLSDP.

This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDE
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8O8F3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IDE
Gene Accession No. P14735, P35559, Q9JHR7
Gene Alias 1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603
Gene Symbols IDE
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Insulin-degrading enzyme (IDE) (P14735).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15925, 25700, 3416
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.