Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDE Monoclonal Antibody (GT143)
GREENER_CHOICE

Catalog No. PIMA527757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527757 Supplier Invitrogen™ Supplier No. MA527757
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: mouse brain, rat brain, HeLa, A431 Predicted Reactivity: Zebrafish (90%), Dog (99%), Rhesus Monkey (100%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GT143
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IDE
Gene Accession No. P14735, P35559, Q9JHR7
Gene Alias 1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603
Gene Symbols IDE
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human IDE. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15925, 25700, 3416
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.