Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDE Monoclonal Antibody (GT286)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527758
Description
Positive Control: HeLa, A431, mouse brain, rat brain Predicted Reactivity: Zebrafish (90%), Dog (99%), Rhesus Monkey (100%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
Specifications
IDE | |
Monoclonal | |
1 mg/mL | |
PBS with 20% glycerol and no preservative; pH 7 | |
P14735, P35559, Q9JHR7 | |
IDE | |
Recombinant protein encompassing a sequence within the N-terminus region of human IDE. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
GT286 | |
Unconjugated | |
IDE | |
1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603 | |
Mouse | |
Protein G | |
RUO | |
15925, 25700, 3416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction