Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDE Monoclonal Antibody (OTI4E5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527528
Description
IDE Monoclonal Antibody for Western Blot
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
Specifications
IDE | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14735, P35559 | |
IDE | |
Human recombinant protein fragment corresponding to amino acids 800-1019 of IDE produced in E.coli. | |
100 μL | |
Primary | |
Human, Rat, Monkey | |
Antibody | |
IgG2b |
Western Blot | |
OTI4E5 | |
Unconjugated | |
IDE | |
1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603 | |
Mouse | |
Affinity Chromatography | |
RUO | |
25700, 3416 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction